missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 beta Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99635CL1
This item is not returnable.
View return policy
Description
14-3-3 beta Polyclonal antibody specifically detects 14-3-3 beta in Human samples. It is validated for ELISA,Immunoprecipitation,Western Blot
Specifications
| 14-3-3 beta | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 7529 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Immunoprecipitation, Western Blot | |
| CoraFluor 1 | |
| 14-3-3 alpha, 14-3-3 beta, 14-3-3 protein beta/alpha | |
| Produced in rabbits immunized with purified, recombinant Human 14-3-3 beta (Accession#: NP_003395.1; Met1-Asn246). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction