missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 epsilon Antibody (02), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-26078MFV610
This item is not returnable.
View return policy
Description
14-3-3 epsilon Monoclonal antibody specifically detects 14-3-3 epsilon in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| 14-3-3 epsilon | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| 02 | |
| Western Blot, Immunoprecipitation | |
| 14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide | |
| Hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human 14-3-3 epsilon (Accession#: NP_006752.1, Met1-Gln255) | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism | |
| 7531 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction