missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 epsilon Antibody (CPTC-YWHAE-1), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-79944CL1
This item is not returnable.
View return policy
Description
14-3-3 epsilon Monoclonal antibody specifically detects 14-3-3 epsilon in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Protein Array
Specifications
| 14-3-3 epsilon | |
| Monoclonal | |
| CoraFluor 1 | |
| 14-3-3 epsilon, 14-3-3 protein epsilon, FLJ45465, FLJ53559,14-3-3E, KCIP-1, MDCR, MDS, mitochondrial import stimulation factor L subunit, protein kinase C inhibitor protein-1, tyrosine 3/tryptophan 5 -monooxygenase activation protein, epsilon polypeptide, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, epsilonpolypeptide | |
| Recombinant human full-length 14-3-3 epsilon protein (Uniprot: P62268) | |
| 0.1 mL | |
| Cancer, Lipid and Metabolism | |
| 7531 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a κ |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Protein Array | |
| CPTC-YWHAE-1 | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction