missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 eta Antibody (3G12), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-92691CL1
This item is not returnable.
View return policy
Description
14-3-3 eta Monoclonal antibody specifically detects 14-3-3 eta in Bovine,Canine,Equine,Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Western Blot,Gene Knock-Out,Immunocytochemistry/Immunofluorescence
Specifications
| 14-3-3 eta | |
| Monoclonal | |
| CoraFluor 1 | |
| 14-3-3 protein eta, Protein AS1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, etapolypeptide, YWHA114-3-3 eta | |
| Full length recombinant human 14-3-3 eta expressed in and purified from E.coli [UniProt# Q04917]. | |
| 0.1 mL | |
| Apoptosis, Cancer, Neuroscience | |
| 7533 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Gene Knock-Out, Immunocytochemistry/Immunofluorescence | |
| 3G12 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Bovine, Canine, Equine, Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction