missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 14-3-3 eta Monoclonal Antibody (06)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA529648
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human 14-3-3 eta/YWHAH.
This gene product belongs to the 14-3-3 family of proteins which mediate signal transduction by binding to phosphoserine-containing proteins. This highly conserved protein family is found in both plants and mammals, and this protein is 99% identical to the mouse, rat and bovine orthologs. This gene contains a 7 bp repeat sequence in its 5' UTR, and changes in the number of this repeat has been associated with early-onset schizophrenia.
Specifications
| 14-3-3 eta | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q04917 | |
| YWHAH | |
| Recombinant Human 14-3-3 eta/YWHAH protein (Gly2-Asn246). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot | |
| 06 | |
| Unconjugated | |
| YWHAH | |
| 14-3-3 eta; 14-3-3 protein eta; 14-3-3 protein eta-subtype; 14-3-3e; 1433F; fb93e02; Protein AS1; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein eta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta; tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, eta polypeptide; wu:fb51h04; wu:fb93e02; YWHA1; YWHA114-3-3 eta; ywhag; YWHAH; zgc:63883; zgc:63883 protein | |
| Mouse | |
| Protein A | |
| RUO | |
| 7533 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction