missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 gamma, ac Val2 Antibody (KC21), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-406F
This item is not returnable.
View return policy
Description
14-3-3 gamma Monoclonal specifically detects 14-3-3 gamma in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| 14-3-3 gamma (ac Val2) | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| YWHAG | |
| N-terminal fragment of human 14-3-3 gamma where the N-terminal Val was acetylated [UniProt# P61981] | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| This antibody is specific for the naturally occurring human form of 14-3-3 gamma, where the N-terminal Met is removed, resulting in an acetylated N-terminal Val. Unprocessed (non-modified) 14-3-3 gamma is not recognized by this antibody. | |
| Store at 4°C in the dark. |
| Immunocytochemistry, Immunofluorescence | |
| KC21 | |
| Immunocytochemistry/Immunofluorescence | |
| 14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction