missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 gamma, ac Val2 Antibody (KC21), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-406MFV500
This item is not returnable.
View return policy
Description
14-3-3 gamma Monoclonal antibody specifically detects 14-3-3 gamma in Human samples. It is validated for Western Blot, Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| 14-3-3 gamma | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Simple Western, Immunocytochemistry | |
| KC21 | |
| Western Blot, Simple Western, Immunocytochemistry/ Immunofluorescence | |
| 14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide | |
| N-terminal fragment of human 14-3-3 gamma where the N-terminal Val was acetylated [UniProt# P61981] | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 7532 | |
| Store at 4C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction