missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 gamma Antibody (AT4B9), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50579AF532
This item is not returnable.
View return policy
Description
14-3-3 gamma Monoclonal specifically detects 14-3-3 gamma in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| 14-3-3 gamma | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| YWHAG | |
| Recombinant human 14-3-3 gamma (1-247aa) purified from E. coli | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 κ |
| Western Blot, ELISA, Immunocytochemistry, Immunofluorescence | |
| AT4B9 | |
| Western Blot, ELISA, Immunocytochemistry/Immunofluorescence | |
| 14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide | |
| Mouse | |
| Protein A purified | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 7532.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction