missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 gamma Antibody (S01-4C7), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20083-100ul
This item is not returnable.
View return policy
Description
14-3-3 gamma Monoclonal antibody specifically detects 14-3-3 gamma in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| 14-3-3 gamma | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S01-4C7 | |
| Unconjugated | |
| 14-3-3 gamma, 14-3-3 protein gamma, 14-3-3GAMMA, KCIP-1, Protein kinase C inhibitor protein 1, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, gammapolypeptide | |
| A synthetic peptide of human 14-3-3 gamma (Uniprot # P61981) | |
| 100 μg | |
| Cancer, Cell Cycle and Replication, Cellular Markers, Growth and Development, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers | |
| 7532 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction