missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 sigma/Stratifin Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99629-100ul
This item is not returnable.
View return policy
Description
14-3-3 sigma/Stratifin Polyclonal antibody specifically detects 14-3-3 sigma/Stratifin in Human samples. It is validated for ELISA
Specifications
| 14-3-3 sigma/Stratifin | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| 14-3-3 protein sigma, 14-3-3 sigma, Epithelial cell marker protein 1, HME1, stratifin, YWHAS | |
| Produced in rabbits immunized with purified, recombinant Human 14-3-3 sigma/Stratifin (Accession#: NP_006133.1; Met 1-Ser 248) | |
| 100 μg | |
| Cell Cycle and Replication | |
| 2810 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction