missing translation for 'onlineSavingsMsg'
Learn More

14-3-3 tau/theta Antibody (1A1), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-21301IR

Product Code. 18420143

  • € 527.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

14-3-3 tau/theta Monoclonal antibody specifically detects 14-3-3 tau/theta in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

14-3-3 tau/theta
Monoclonal
DyLight 755
14-3-3 protein tau, 14-3-3 protein T-cell, 14-3-3 protein theta, 14-3-3,1C5, HS1, Protein HS1, protein tau, tyrosine 3-monooxygenase/tryptophan 5-monooxygenase activation protein, thetapolypeptide
Recombinant human 14-3-3 tau/theta (1-245aa) purified from E. coli (NP_006817).
0.1 mL
Apoptosis, Cell Biology, Cell Cycle and Replication, Neuroscience, Signal Transduction, Stem Cells
10971
Store at 4°C in the dark.
IgG1 κ
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
1A1
50mM Sodium Borate
Mouse
Protein G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.