missing translation for 'onlineSavingsMsg'
Learn More
Learn More
14-3-3 theta Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538397
This item is not returnable.
View return policy
Description
At least seven isoforms comprise the highly conserved 14-3-3 family of homo- and heterodimeric proteins that are abundantly expressed in all eukaryotic cells. Although more than seven isoforms of 14-3-3 have been described, some redundancies have appeared upon sequencing. The 14-3-3s are thought to be key regulators of signal transduction events mediated through their binding to serine-phosphorylated proteins. By interacting with Cdc25C, 14-3-3 regulates entry into the cell cycle and, through interaction with Bad, prevents apoptosis. Other proteins that have been shown to bind to 14-3-3s include members of the protein kinase C family, Cbl, IRS-1, polyoma middle-T antigen, nitrate reductase, S-raf and the IGF-1 receptor. Detection of 14-3-3 proteins in cerebrospinal fluid has been shown to be quite useful in the differential diagnosis of Creutzfeldt-Jakob disease and other prion diseases.
Specifications
| 14-3-3 theta | |
| Polyclonal | |
| Unconjugated | |
| YWHAQ | |
| 14-3-3, 1C5, HS1, 14-3-3 protein T-cell, 14-3-3 protein tau, 14-3-3 protein theta, protein tau | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 10971, 22630 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 1 mg/mL | |
| Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| P27348, P68254 | |
| YWHAQ | |
| A synthetic non-phosphopeptide derived from human 14-3-3 theta/tau around the phosphorylation site of Ser232 (S-D-SP-A-G) | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction