Learn More
Keystone Cleanroom Products 1422A Tyvek™ Zipper Equipment Cover
GREENER_CHOICE
Product Code. 17708307 Shop All Keystone Cleanroom Products Products
Click to view available options
Length (English):
26 in.
30 in.
32 in.
50 in.
54 in.
Width (English):
22 in.
24 in.
26 in.
28 in.
54 in.
Height (English):
44 in.
50 in.
56 in.
72 in.
80 in.
Unit Size:
Pack of 50
Product Code. 17708307
Brand: Keystone Cleanroom Products COVTVK502872Z2WP4
Only null left
Add to basket
Add to basket
1422A Tyvek™ Zipper Equipment Cover is an ideal choice in pharmaceutical operations to help keep clean equipment protected from both environmental and product cross contamination.
  • Keeps Cleaned Equipment Clean and Protected from Contamination
  • Breathable
  • Form Fitting, Fast and Consistent
  • Manufactured and packaged in a ISO 6 cleanroom
  • Conveniently packaged: 1 heat-sealed labeled poly bag
  • Certificates of Conformance sent with shipment
Specifications
Autoclavable Autoclavable
Certifications/Compliance COC
Closure Type Zip Closure
Color White
Disposable Single-use
For Use With (Application) Steam Sterilization
For Use With (Equipment) Pharmaceutical Equipment
Material Tyvek
Sterility Non-sterile
Latex Free Yes
Sterilization Method Steam
Temperature Range (English) <250°F
Temperature Range (Metric) <125°C
Dimensions (L x W x H) 50 x 28 x 72 in.
Length (English) 50 in.
Length (Metric) 127 cm
Width (English) 28 in.
Width (Metric) 71 cm
Height (English) 72 in.
Height (Metric) 183 cm
Product Type Cover