Learn More
15α-Acetoxy-gestodene, TRC
Specifications
Specifications
| Chemical Name or Material | 13-Ethyl-17-hydroxy-3-oxo-18,19-dinor-17Alpha-pregn-4-en-20-yn-15Alpha-yl Acetate |
| CAS | 267650-77-1 |
| Molecular Formula | C23 H30 O4 |
| Quantity | 50 mg |
| Synonym | 18,19-Dinorpregn-4-en-20-yn-3-one, 15-(acetyloxy)-13-ethyl-17-hydroxy-, (15α,17α)- (9CI), 13-Ethyl-17-hydroxy-3-oxo-18,19-dinor-17α-pregn-4-en-20-yn-15α-yl acetate, 15α-Acetoxy-gestodene, Gestodene Impurity F (EP) |
| InChI Formula | InChI=1S/C23H30O4/c1-4-22-11-10-18-17-9-7-16(25)12-15(17)6-8-19(18)21(22)20(27-14(3)24)13-23(22,26)5-2/h2,12,17-21,26H,4,6-11,13H2,1,3H3/t17-,18+,19+,20-,21+,22-,23-/m0/s1 |
| SMILES | CC[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)CC[C@H]34)[C@@H]1[C@H](C[C@@]2(O)C#C)OC(=O)C |
| IUPAC Name | [(8R,9S,10R,13S,14S,15S,17R)-13-ethyl-17-ethynyl-17-hydroxy-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-15-yl] acetate |
| Molecular Weight (g/mol) | 370.482 |
| Formula Weight | 370.214 |