Learn More
15-PGDH/HPGD Antibody (016), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-89839H
Product Code. 18656601
  • € 607.00 / 0.10mL
Description
15-PGDH/HPGD Monoclonal antibody specifically detects 15-PGDH/HPGD in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
HRP
Cancer, Immunology, Innate Immunity