missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 15-PGDH Recombinant Rabbit Monoclonal Antibody (025), HRP

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552289
This item is not returnable.
View return policy
Description
15-PGDH Recombinant Monoclonal Antibody for ELISA
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.
Specifications
| 15-PGDH | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with no preservative | |
| P15428 | |
| HPGD | |
| Recombinant Human HPGD/15-PGDH protein (Met1-Gln266). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| ELISA | |
| 025 | |
| HRP | |
| HPGD | |
| 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NAD(+)]; 15-PGDH; AV026552; HPGD; hydroxyprostaglandin dehydrogenase 15 (NAD); hydroxyprostaglandin dehydrogenase 15-(NAD); NAD+-dependent 15-hydroxyprostaglandin dehydrogenase; NAD-dependent 15-hydroxyprostaglandin dehydrogenase; PGDH; PGDH1; PHOAR1; Prostaglandin dehydrogenase 1; SDR36C1; Short chain dehydrogenase/reductase family 36C member 1; short chain dehydrogenase/reductase family 36C, member 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3248 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction