Learn More
Invitrogen™ 15-PGDH Recombinant Rabbit Monoclonal Antibody (101)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA529714
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Mouse HPGD/15-PGDH.
Prostaglandin inactivation. Contributes to the regulation of events that are under the control of prostaglandin levels. Catalyzes the NAD-dependent dehydrogenation of lipoxin A4 to form 15-oxo-lipoxin A4. Inhibits in vivo proliferation of colon cancer cells.
Specifications
| 15-PGDH | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| PBS with no preservative | |
| Q8VCC1 | |
| HPGD | |
| Recombinant Mouse HPGD/15-PGDH protein (Met1-Ser269). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
| Immunoprecipitation, Western Blot | |
| 101 | |
| Unconjugated | |
| HPGD | |
| 15-hydroxyprostaglandin dehydrogenase; 15-hydroxyprostaglandin dehydrogenase [NAD(+)]; 15-PGDH; AV026552; HPGD; hydroxyprostaglandin dehydrogenase 15 (NAD); hydroxyprostaglandin dehydrogenase 15-(NAD); NAD+-dependent 15-hydroxyprostaglandin dehydrogenase; NAD-dependent 15-hydroxyprostaglandin dehydrogenase; PGDH; PGDH1; PHOAR1; Prostaglandin dehydrogenase 1; SDR36C1; Short chain dehydrogenase/reductase family 36C member 1; short chain dehydrogenase/reductase family 36C, member 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 15446 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.