Learn More
Hamilton™ 16 gauge, Hubless Needle, custom length (0.375 to 12 in), point style 5 or AS, 6/PK
Product Code. 12351798
Click to view available options
Needle Gauge:
16 G
Unit Size:
Pack of 6
Product Code. 12351798
Brand: Hamilton™ 2201602
This category of needles comes without a hub so they cannot be connected to a Hamilton syringe. The point style and length of the tubing can be specified at the time of order.
Specification Sheet
  • Gauge – 16 gauge
  • Point Style – 5 or AS
  • Needle Length – Custom Needle Length
  • Autoclavable – Yes
  • Sales Quantity – 6 needles per pack
  • Hub – No Hub
Specifications
Needle Gauge 16 G
Product Type Hubless Needles