Learn More
16-Hydroxyhexadecanoic acid, 97%
Product Code. 11315139
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11315139 1 g 1g
11325139 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11315139 Supplier Thermo Scientific Alfa Aesar Supplier No. A17845.03
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
missing translation for 'validMainframeMsg'
CAS: 506-13-8 | C16H32O3 | 272.429 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 506-13-8
Molecular Formula C16H32O3
Molecular Weight (g/mol) 272.429
MDL Number MFCD00002750
InChI Key UGAGPNKCDRTDHP-UHFFFAOYSA-N
Synonym juniperic acid, 16-hydroxypalmitic acid, hexadecanoic acid, 16-hydroxy, palmitic acid, 16-hydroxy, omega-hydroxypalmitic acid, 16-hydroxy-hexadecanoic acid, unii-7ipp3u0f3i, .omega.-hydroxypalmitic acid, 7ipp3u0f3i, 16-hydroxy hexadecanoic acid
PubChem CID 10466
ChEBI CHEBI:55328
IUPAC Name 16-hydroxyhexadecanoic acid
SMILES C(CCCCCCCC(=O)O)CCCCCCCO
Tekniske data
Melting Point 98°C to 100°C
Linear Formula HO(CH2)15CO2H
Quantity 1 g
Beilstein 1783998
Formula Weight 272.43
Percent Purity 98%
Chemical Name or Material 16-Hydroxyhexadecanoic acid
RUO – Research Use Only
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.