Learn More
Thermo Scientific™ 16 mL Nalgene Oak Ridge Tube Adapters for Fiberlite F13-14x50cy Rotor and F15-8x50cy Rotor
Product Code. 15382167
Product Code. 15382167
missing translation for 'mfr': Thermo Scientific™ 0100376
Accommodate 16mL Oak Ridge tubes in Thermo Scientific™ Fiberlite™ conical tube rotors.
Accommodate 16mL Nalgene Oak Ridge tubes in Thermo Scientific™ Fiberlite™ F13-14x50cy or Fiberlite F15-8x50cy rotors.
TRUSTED_SUSTAINABILITY
Specifications
Type Ridge Tube Adapter
Tube Type 16 mL Nalgene Oak Ridge Tube
Capacity 16 mL
For Use With (Equipment) Fiberlite F13-14x50cy, Fiberlite F15-8x50cy rotors
Description 16mL Nalgene Oak Ridge Tube Adapter