missing translation for 'onlineSavingsMsg'
Learn More

17-Desacetyl Norgestimate-d6 (Major), TRC

Product Code. 30365999
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365999 1 mg 1mg
30357303 25 mg 25mg
30356073 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30365999 Supplier Toronto Research Chemicals Supplier No. TRCD2887021MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1263184-13-9
Molecular Formula C21 D6 H23 N O2
Molecular Weight (g/mol) 333.497
Synonym 18,19-Dinorpregn-4-en-20-yn-3-one-2,2,4,6,6,10-d6, 13-ethyl-17-hydroxy-, oxime, (17α)-, Deacetyl norgestimate-d6, 17-Desacetyl norgestimate-d6, Norelgestromin-d6, Norelgestromin-2,2,4,6,6,10-D6, (17α)-13-Ethyl-17-hydroxy-18,19-dinorpregn-4-en-20-yn-3-one-2,2,4,6,6,10-d6 oxime, 17-Deacetylnorgestimate-2,2,4,6,6,10-d6, Desacetylnorgestimate-d6
IUPAC Name (8R,9S,10R,13S,14S,17R)-2,2,4,6,6,10-hexadeuterio-13-ethyl-17-ethynyl-3-hydroxyimino-7,8,9,11,12,14,15,16-octahydro-1H-cyclopenta[a]phenanthren-17-ol
SMILES [2H]C1=C2C([2H])([2H])C[C@@H]3[C@H](CC[C@@]4(CC)[C@H]3CC[C@@]4(O)C#C)[C@@]2([2H])CC([2H])([2H])C1=NO

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C21H29NO2/c1-3-20-11-9-17-16-8-6-15(22-24)13-14(16)5-7-18(17)19(20)10-12-21(20,23)4-2/h2,13,16-19,23-24H,3,5-12H2,1H3/t16-,17+,18+,19-,20-,21-/m0/s1/i5D2,6D2,13D,16D
Formula Weight 333.257
Chemical Name or Material 17-Desacetyl Norgestimate-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.