missing translation for 'onlineSavingsMsg'
Learn More

17-Northebaine, Mikromol™

Product Code. 18093300
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18093300 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18093300 Supplier Mikromol Supplier No. MM0244.020025

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

17-Northebaine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 17-Northebaine
CAS 2579-67-1
Analyte or Component Names 17-Northebaine
Shipping Condition Room Temperature
Molecular Formula C18 H19 N O3
Certifications/Compliance ISO 17025
Synonym Morphinan, 6,7,8,14-tetradehydro-4,5α-epoxy-3,6-dimethoxy- (8CI), Northebaine (7CI), (5α)-6,7,8,14-Tetradehydro-4,5-epoxy-3,6-dimethoxymorphinan, (5α)-6,7,8,14-tetradehydro-4,5-epoxy-3,6-dimethoxy-morphinan, (-)-Northebaine, N-Demethylthebaine, N-Northebaine
InChI Formula InChI=1S/C18H19NO3/c1-20-13-5-3-10-9-12-11-4-6-14(21-2)17-18(11,7-8-19-12)15(10)16(13)22-17/h3-6,12,17,19H,7-9H2,1-2H3/t12-,17+,18+/m1/s1
SMILES COC1=CC=C2[C@H]3Cc4ccc(OC)c5O[C@@H]1[C@]2(CCN3)c45
IUPAC Name (4R,7aR,12bS)-7,9-dimethoxy-1,2,3,4,7a,13-hexahydro-4,12-methanobenzofuro[3,2-e]isoquinoline
Molecular Weight (g/mol) 297.35
Quantity 25 mg
Formula Weight 297.1365 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.