Learn More
Hamilton™ Gel-Loading 8 Channel Removable Needle Replacements
Product Code. 12660343
Product Code. 12660343
Brand: Hamilton™ 84711
Designed for multiplex DNA sequencing methodologies.
  • Rapid sample transfer
  • Fixed, 25mm long, blunt needles spaced 9mm apart
  • Adjustable volume stop allows faster, more accurate reloading and excellent reproducibility