Learn More
Bochem™ 18/10 Stainless Steel Chemical Scoop
Click to view available options
Capacity (Metric):
450 mL
Unit Size:
Each
Description
- 18/10 Stainless Steel
Specifications
Specifications
| Quantity | 1 Ea. |
| Capacity (Metric) | 450 mL |
| Product Type | Chemical Scoop |
| For Use With (Application) | Manual scoops for collecting or measuring powders, pastes or granulates |
| Material | Stainless Steel (18/10) |
| Length (Metric) | 250 mm |