Learn More
Bochem™ 18/10 Stainless Steel Coverglass Forceps
Product Code. 10241711
Click to view available options
Tip Style:
Bent
Straight
Unit Size:
Each
Pack of 10
Product Code. 10241711
Brand: Bochem™ 1727
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from 18/10 stainless steel. Bochem™ 18/10 Stainless Steel Coverglass Forceps has 6mm wide points.
  • Coverglas forceps
  • Stainless steel
  • Points 6mm wide
Specifications
Tip Style Straight
Material Stainless Steel (18/10)
Length (Metric) 105 mm
Product Type Forceps
Quantity (No. per Case) 10/Pack
For Use With (Application) Good grip risk-free membranes, autoclaving or flame, pliers
Type Coverglass Forceps