Learn More
Bochem™ 18/10 Stainless Steel Cross Type Boss heads
Certified DIN 12895. Bochem™ 18/10 Stainless Steel Cross Type Boss heads comes in three types.
Brand: Bochem™ 5306
Additional Details : Weight : 0.10000kg
Description
- 18/10 stainless steel
- DIN 12895
- Three types: Heart screw M6; Socket screw M6; and safety screw type "heart" M8
Specifications
Stainless Steel (18/10) | |
Cross Type Boss head | |
8 mm | |
1 Pk. |