Learn More
Bochem™ 18/10 Stainless Steel Double Ended Micro-spatulas
Click to view available options
Length (Metric):
100 mm
130 mm
150 mm
185 mm
210 mm
Length (Metric) Blade:
30 mm
35 mm
40 mm
50 mm
55 mm
Width (Metric) Blade:
2 mm
3 mm
4 mm
5 mm
6 mm
9 mm
Unit Size:
Each
Beskrivelse
- Double endet
- Stainless steel
- Autoclavable
- Straight
Tekniske data
Tekniske data
| Quantity | 1 Ea. |
| Autoclavable | Autoclavable |
| Shape | Straight |
| Product Type | Double Ended Micro Spatula |
| For Use With (Application) | Multi-purpose spatula for a wide range of uses |
| Length (Metric) Blade | 40 mm |
| Width (Metric) | 2 mm |
| Width (Metric) Blade | 2 mm |
| Blade Material | Stainless Steel (18/10) |
| Diameter (Metric) Outer | 1mm |
| Vis mere |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion