Learn More
Bochem™ 18/10 Stainless Steel Pharma Scoops
Product Code. 12621895
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Each
Product Code. 12621895
Brand: Bochem™ 8835
Only null left
Add to basket
Add to basket
Includes hanging loop. Bochem 18/10 Stainless Steel Pharma Scoops are qualified for food handling.
  • Seamless fixed handle, extremly high processing
  • With hanging loop
  • Qualified for food
Specifications
Capacity (Metric) 100 mL
Product Type Pharma Scoop
For Use With (Application) Qualified for food
Width (Metric) 60 mm
Dimensions (L x W) 215 x 60 mm
Includes With Hanging Loop
Material Stainless Steel (18/10)
Handle Type Seamless fixed handle
Length (Metric) 215 mm