Learn More
Bochem™ 18/10 Stainless Steel Poly-Spoon
Product Code. 10166950
Click to view available options
Length (Metric):
210 mm
Unit Size:
Each
Product Code. 10166950
Brand: Bochem™ 3402
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Made from 18/10 Stainless Steel.
  • Type 1
  • Stainless Steel
Specifications
Shape Bent
Product Type Poly Spoon
Width (Metric) 13 mm
Width (Metric) Blade 13 mm
Material Stainless Steel (18/10)
Length (Metric) 210 mm