Learn More
Bochem™ 18/10 Stainless Steel Propeller Stirrer
Product Code. 15256427
Click to view available options
Diameter (Metric):
70 mm
100 mm
Unit Size:
Each
Product Code. 15256427
Brand: Bochem™ 9012
Only null left
Specifications
Description Propeller stirrer, 4 fix blades
Diameter (Metric) 70 mm
Diameter (Metric) Shaft 8 mm
For Use With (Application) Mixing of media with low and middle viscosity
Length (Metric) 500 mm