Learn More
Bochem™ 18/10 Stainless Steel Section Lifters, with Wooden Handle
Manufactured from 18/10 stainless steel
Brand: Bochem™ 3535
Additional Details : Weight : 0.00900kg
Specifications
1 Ea. | |
50 mm | |
Stainless Steel (18/10) | |
150 x 10 mm | |
Stainless Steel (18/10) |
Section Lifter | |
10 mm | |
0.4mm | |
Wood | |
150 mm |