missing translation for 'onlineSavingsMsg'
Learn More

19-Carboxy Cholesterol, TRC

Product Code. 30361772
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30361772 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30361772 Supplier Toronto Research Chemicals Supplier No. TRCC1778455MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 19-Carboxy Cholesterol
CAS 26319-96-0
Molecular Formula C28 H46 O3
Quantity 5 mg
Synonym (3beta)-3-Hydroxycholest-5-en-19-oic Acid
InChI Formula InChI=1S/C28H46O3/c1-18(2)6-5-7-19(3)23-10-11-24-22-9-8-20-16-21(29)12-15-28(20,17-26(30)31)25(22)13-14-27(23,24)4/h8,18-19,21-25,29H,5-7,9-17H2,1-4H3,(H,30,31)/t19-,21+,22+,23-,24+,25+,27-,28-/m1/s1
SMILES CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(CC(=O)O)[C@H]3CC[C@]12C
IUPAC Name 2-[(3S,8S,9S,10S,13R,14S,17R)-3-hydroxy-13-methyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-10-yl]acetic acid
Molecular Weight (g/mol) 430.663
Formula Weight 430.345
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.