Learn More
Paragon Scientific 190GR Vapour Pressure ASTM D5191: DVPE..@ 66.7 kPa
Brand:  Paragon Scientific ALK-CRMU-VP05
190GR Vapour Pressure ASTM D5191: DVPE..@ 66.7 kPa Ptot ..73.1 kPa
Product Code. 30458382
  • € 744.00 / 190g