missing translation for 'onlineSavingsMsg'
Learn More
Learn More
V-Solv™ ICP Solvent, VHG™
Click to view available options
Quantity:
1 gal
5 gal
200 L
Unit Size:
1 GAL
200 L
5 GAL
Description
V-Solv™ ICP Solvent
Specifications
Specifications
| Methods | ICP |
| Shipping Condition | Room Temperature |
| Solvent or Matrix | V-SOLV™ ICP Solvent |
| Quantity | 1 gal |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction