missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,3-Butane-1,1,1,4,4,4-d6-diol (mixture of stereoisomers), CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | 2,3-Butanediol-1,1,1,4,4,4-D6 |
| CAS | 344750-80-7 |
| Molecular Formula | C4 D6 H4 O2 |
| Quantity | 1 g |
| Synonym | 2,3-Butane-1,1,1,4,4,4-d6-diol (9 CI), 2,3-Butanediol-d6, 2,3-Butanediol-1,1,1,4,4,4-D6, 2,3-Butanediol-1,1,1,4,4,4-d6 |
| InChI Formula | InChI=1 S/C4H10O2/c1-3(5)4(2)6/h3-6 H,1-2H3/i1D3,2D3 |
| SMILES | [2 H]C([2 H])([2 H])C(O)C(O)C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,4,4,4-hexadeuteriobutane-2,3-diol |
| Molecular Weight (g/mol) | 96.158 |
| Formula Weight | 96.1057 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction