missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(2-pyridyl)-1-phenyl-1,2-dihydropyridin-2-one, TRC

Product Code. 30341928
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30341928

Brand: Toronto Research Chemicals TRCB6872501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromo-5-(2-pyridyl)-1-phenyl-1,2-dihydropyridin-2-one
CAS 381248-06-2
Molecular Formula C16H11BrN2O
Quantity 1 g
Synonym 3-Bromo-1-phenyl-5-(pyridin-2-yl)-2(1H)-pyridone, 5'-Bromo-1'-phenyl-[2,3'-bipyridin]-6'(1'H)-one
InChI Formula InChI=1S/C16H11BrN2O/c17-14-10-12(15-8-4-5-9-18-15)11-19(16(14)20)13-6-2-1-3-7-13/h1-11H
SMILES BrC1=CC(=CN(C1=O)c2ccccc2)c3ccccn3
IUPAC Name 3-bromo-1-phenyl-5-pyridin-2-ylpyridin-2-one
Molecular Weight (g/mol) 327.18
Formula Weight 326.0055
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.