Learn More
3-Methylbutyryl-d9 Chloride, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Specifications
Specifications
| Chemical Name or Material | 3-Methylbutanoyl Chloride-D9 |
| CAS | 1219803-46-9 |
| Molecular Formula | C5 D9 Cl O |
| Quantity | 1 g |
| Synonym | 3-(Methyl-d3)butanoyl-2,2,3,4,4,4-d6 chloride, Isovaleryl chloride-d9, 3-Methylbutyryl chloride-d9, 3-Methylbutyryl-d9 Chloride, 3-Methylbutanoyl chloride-d9, 3-Methylbutanoyl Chloride-D9 |
| InChI Formula | InChI=1 S/C5H9ClO/c1-4(2)3-5(6)7/h4H,3H2,1-2H3/i1D3,2D3,3D2,4 D |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])C(=O)Cl |
| IUPAC Name | 2,2,3,4,4,4-hexadeuterio-3-(trideuteriomethyl)butanoyl chloride |
| Molecular Weight (g/mol) | 129.6328 |
| Formula Weight | 129.0907 g/mol |
| Show More |