Learn More
Benzyl-2,3,4,5,6-d5 Bromide, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Benzyl-d5 Bromide |
| CAS | 71258-22-5 |
| Molecular Formula | C7 2H5 H2 Br |
| Quantity | 1 g |
| InChI Formula | InChI=1 S/C7H7Br/c8-6-7-4-2-1-3-5-7/h1-5 H,6H2/i1D,2 D,3 D,4 D,5 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(CBr)c([2 H])c1[2 H] |
| IUPAC Name | 1-(bromomethyl)-2,3,4,5,6-pentadeuteriobenzene |
| Molecular Weight (g/mol) | 176.07 |
| Formula Weight | 175.0045 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |