Learn More
Ethylene-d4-diamine 2HCl, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Ethylenediamine-1,1,2,2-D4 Dihydrochloride |
| CAS | 34334-71-9 |
| Molecular Formula | C2 2H4 H4 N2 . 2 Cl H |
| Quantity | 1 g |
| Synonym | 1,2-Ethane-1,1,2,2-d4-diamine, dihydrochloride (9 CI), Ethylene-d4-diamine, dihydrochloride (8 CI), 1,2-Ethane-1,1,2,2-d4-diamine dihydrochloride, Ethylene-d4 diamine dihydrochloride, 1,2-Diamino-1,1,2,2-tetradeuteroethane dihydrochloride, Ethylenediamine- |
| InChI Formula | InChI=1 S/C2H8N2.2 ClH/c3-1-2-4;;/h1-4H2;2*1 H/i1D2,2D2;; |
| SMILES | Cl.Cl.[2 H]C([2 H])(N)C([2 H])([2 H])N |
| IUPAC Name | 1,1,2,2-tetradeuterioethane-1,2-diamine;dihydrochloride |
| Molecular Weight (g/mol) | 137.045 |
| Formula Weight | 136.047 g/mol |
| Show More |