missing translation for 'onlineSavingsMsg'
Learn More

TdB Labs AB 400 kDa FITC-polysucrose Green Fluorophore

Product Code. 30216475
Encompass_Preferred
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
This item is not returnable. View return policy

Product Code. 30216475

Brand: TdB Labs AB FP400_1g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

FITC-polysucrose 400 kDa is a high molecular weight, green-fluorescent tracer used in macromolecular diffusion, extracellular volume, and retention studies. Ideal for modeling transport barriers in complex biological systems.

  • Product Overview: FITC-polysucrose is a high molecular weight, water-soluble fluorescent tracer based on polysucrose labeled with fluorescein isothiocyanate (FITC). Offering bright green fluorescence and excellent biocompatibility, FITC-polysucrose is ideal for studies involving permeability, molecular diffusion, vascular barrier function, and biological transport models. Its stability and non-toxicity make it suitable for both live-cell experiments and tissue models.
  • Features:
    • Green Fluorescence: Excitation maximum at 495 nm and emission maximum at 519 nm, compatible with standard FITC filter sets.
    • High Molecular Weight Options: Available in different sizes (e.g., 20 kDa, 40 kDa, 70 kDa, 100 kDa, 400 kDa) to match diffusion and permeability requirements.
    • Water-Soluble and Biocompatible: Fully soluble in aqueous solutions, suitable for biological systems without inducing cytotoxicity.
    • Bright and Stable Labeling: FITC conjugation provides durable fluorescence for both real-time tracking and fixed endpoint studies.
    • Minimal Non-Specific Binding: Maintains clear background and reliable signal detection in complex biological environments.
  • Benefits:
    • Reliable Molecular Diffusion Tracking: Models macromolecule behavior across biological barriers and in engineered systems.
    • Ideal for Permeability Assays: Commonly used to assess tight junction integrity, vascular leakage, and epithelial barrier function.
    • Supports Live-Cell and Fixed-Cell Imaging: Suitable for both dynamic real-time experiments and post-fixation endpoint analysis.
    • Flexible Use in Diverse Systems: Applied in microfluidic platforms, organ-on-chip models, perfused tissues, and 3D constructs.
    • Bright Fluorescence for Easy Quantification: Ensures sensitive and reproducible detection across imaging and plate reader platforms.
  • Applications:
    • Permeability and Barrier Integrity Studies: Used in epithelial, endothelial, and blood-brain barrier models to assess transport properties.
    • Tracer Studies in Microfluidics and Bioreactors: Visualizes molecular transport in flow-based systems.
    • Live and Fixed Imaging Applications: Tracks distribution of macromolecules in live-cell or fixed-tissue experiments.
    • Diffusion and Transport Modeling: Supports research into size-dependent molecular movement in biological and synthetic systems.
  • Technical Specifications:
    • Fluorescence: Excitation maximum at 495 nm, emission maximum at 519 nm.
    • Molecular Weights: Available in multiple sizes, including 20 kDa, 40 kDa, 70 kDa, 100 kDa, and 400 kDa.
    • Appearance: Light-sensitive, greenish dry powder, readily soluble in water.
    • Labeling: Optimized degree of labeling to ensure bright fluorescence while maintaining polysucrose behavior.
  • Storage and Stability:
    • Store dry and protected from light at 2°C to 8°C for optimal stability and fluorescence retention.
    • Stable under recommended conditions for extended periods without significant loss of performance.
  • Packaging and Availability:
    • Supplied as dry powder to maximize stability and handling ease.
    • Available in multiple research-grade packaging sizes to support both small-scale and larger experimental needs.

Tekniske data

Detection Method Fluorescence
Format Vial
Product Type Reagent
Quantity 1 g
Form Crystalline Powder
Korrektion af produktindhold

Fisher Scientific Encompass-programmet tilbyder varer, som ikke er en del af vores distributionsportefølje. Disse produkter har typisk ikke billeder eller detaljerede beskrivelser. Vi er dog forpligtet til at forbedre din shoppingoplevelse. Brug formularen nedenfor til at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.