Learn More
Methyl-d1 Alcohol, CDN
Product Code. 30309376
Click to view available options
Quantity:
1 g
Unit Size:
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30309376
Brand: CDN CDND04681G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Methanol-d
CAS 4206-31-9
Molecular Formula C D H3 O
Quantity 1 g
Synonym Methanol-d, Methan-d-ol (6 CI, 7 CI, 8 CI, 9 CI), Methanol-d1, Methyl-d alcohol, Methanol-1-D
InChI Formula InChI=1 S/CH4O/c1-2/h2H,1H3/i1D
SMILES [2 H]CO
IUPAC Name deuteriomethanol
Molecular Weight (g/mol) 33.048
Formula Weight 33.0325 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity