Learn More
Methyl-d3 p-Toluenesulfonate, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Methyl-d3 Toluenesulfonate |
| CAS | 7575-93-1 |
| Molecular Formula | C8 D3 H7 O3 S |
| Quantity | 1 g |
| Synonym | Methan-d3-ol, 4-methylbenzenesulfonate (9 CI, ACI), Methan-d3-ol, p-toluenesulfonate (8 CI), p-Toluenesulfonic acid, methyl-d3 ester (6 CI, 7 CI, 8 CI), Methyl-d3 4-methylbenzenesulfonate, Methyl-d3 tosylate, Trideuteriomethyl 4-methoxybenzenesulfonate, Trideuteriomethyl 4-methylbenzenesulfonate, Trideuteriomethyl p-toluenesulfonate, Trideuteromethyl tosylate, Methyl-d3 p-Toluenesulfonate, Methyl Toluene-4-sulfonate-D3 (methoxy-D3) |
| InChI Formula | InChI=1 S/C8H10O3S/c1-7-3-5-8(6-4-7)12(9,10)11-2/h3-6 H,1-2H3/i2D3 |
| SMILES | [2 H]C([2 H])([2 H])OS(=O)(=O)c1ccc(C)cc1 |
| IUPAC Name | trideuteriomethyl 4-methylbenzenesulfonate |
| Molecular Weight (g/mol) | 189.247 |
| Formula Weight | 189.054 g/mol |
| Show More |