missing translation for 'onlineSavingsMsg'
Learn More

Phloroglucide Hydrate, TRC

Product Code. 17982215
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17982215 1 g 1g
30136189 10 g 10g
30136541 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17982215 Supplier Toronto Research Chemicals Supplier No. TRCP3399501G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C12 H10 O5 . x(H2O)
Molecular Weight (g/mol) 234.2
Synonym [1,1'-Biphenyl]-2,3',4,5',6-pentol Hydrate, 2,4,6,3',5'-Biphenylpentol Hydrate, NSC 65069 Hydrate;
IUPAC Name 2-(3,5-dihydroxyphenyl)benzene-1,3,5-triol;hydrate
SMILES OC1=CC(O)=CC(O)=C1C2=CC(O)=CC(O)=C2.O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H10O5.H2O/c13-7-1-6(2-8(14)3-7)12-10(16)4-9(15)5-11(12)17;/h1-5,13-17H;1H2
Chemical Name or Material Phloroglucide Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.