Learn More
Bachem 1GR SASRIN resin (200-400 mesh, 0.80-1.20 mmol/g)
Brand:  Bachem 4012712.0001
Encompass_Preferred
1GR SASRIN resin (200-400 mesh, 0.80-1.20 mmol/g)
Product Code. 16624187
  • € 137.89 / 1g