Learn More
tert-Butyl Alcohol-d10, CDN
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | tert-Butyl Alcohol-d10 |
| CAS | 53001-22-2 |
| Molecular Formula | C4 D10 O |
| Quantity | 1 g |
| Synonym | 2-Propan-1,1,1,3,3,3-d6-ol-d, 2-(methyl-d3)- (9 CI), tert-Butyl-d9 alcohol-d (6 CI, 7 CI), 1,1,1,3,3,3-Hexadeutero-2-deuterooxy-2-(trideuteromethyl)propane, 2-Methyl-2-propanol-d10, tert-Butanol-d10, tert-Butanol-D10, tert-Butyl alcohol-d10, 2-(Methyl-d3)-2-propan-1,1,1,3,3,3-d6-ol-d |
| InChI Formula | InChI=1 S/C4H10O/c1-4(2,3)5/h5H,1-3H3/i1D3,2D3,3D3,5 D |
| SMILES | [2 H]OC(C([2 H])([2 H])[2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,1,1,3,3,3-hexadeuterio-2-deuteriooxy-2-(trideuteriomethyl)propane |
| Molecular Weight (g/mol) | 84.183 |
| Formula Weight | 84.136 g/mol |
| Show More |