missing translation for 'onlineSavingsMsg'
Learn More

1H-Indole-3-acetamide, TRC

Product Code. 30408148
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30408148 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30408148 Supplier Toronto Research Chemicals Supplier No. TRCI5774911G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 879-37-8
Molecular Formula C10H10N2O
Quantity 1 g
Synonym Indole-3-acetamide, 2-(1H-Indol-3-yl)acetamide, 2-(3-Indolyl)acetamide, 3-Indolylacetamide, NSC 1969
InChI Formula InChI=1S/C10H10N2O/c11-10(13)5-7-6-12-9-4-2-1-3-8(7)9/h1-4,6,12H,5H2,(H2,11,13)
SMILES NC(=O)Cc1c[nH]c2ccccc12
IUPAC Name 2-(1H-indol-3-yl)acetamide
Molecular Weight (g/mol) 174.2
Formula Weight 174.0793
Chemical Name or Material 1H-Indole-3-acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.