missing translation for 'onlineSavingsMsg'
Learn More

Sulfur - S @ 0 μg/g (blank) in #2 Diesel Fuel, VHG™

Product Code. 17932276
Click to view available options
Quantity:
100 mL
500 mL
1 L
1 gal
Unit Size:
1 GAL
1 L
100mL
500mL
This item is not returnable. View return policy

Product Code. 17932276

Brand: VHG Labs VHGSDSLBLK1L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Sulfur - S @ 0 μg/g (blank) in #2 Diesel Fuel
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Sulfur
CAS 7704-34-9
Methods XRF
Analyte or Component Names Sulfur
Shipping Condition Room Temperature
Molecular Formula S
InChI Formula InChI=1S/S
SMILES [S]
IUPAC Name sulfur
Solvent or Matrix #2 Diesel Fuel
Molecular Weight (g/mol) 32.065
Quantity 1 L
Formula Weight 31.972 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.