missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 1M7

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
1M7 is a reagent for RNA SHAPE-MaP experiments. Enables analysis of RNA secondary structures at single nucleotide resolution in <70 seconds.
Specifications
Specifications
| Chemical Name or Material | 1-Methyl-7-nitro-2H-3,1-benzoxazine-2,4(1H)-dione |
| CAS | 73043-80-8 |
| Synonym | 1-Methyl-7-nitroisatoic anhydride |
| Quantity | 10 mg |
| Molecular Formula | C9H6N2O5 |
| Formula Weight | 222.15 |
| Purity | 95% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction