missing translation for 'onlineSavingsMsg'
Learn More

1cP-LSD, TRC

Product Code. 30005777
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30005777 1 mg 1mg
30136683 10 mg 10mg
30012489 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30005777 Supplier Toronto Research Chemicals Supplier No. TRCL3550251MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C24H29N3O2
Molecular Weight (g/mol) 391.51
Synonym (6aR,9R)-4-(Cyclopropanecarbonyl)-N,N-diethyl-7-methyl-4,6,6a,7,8,9-hexahydroindolo[4,3-fg]quinoline-9-carboxamide, 1cP-LSD
IUPAC Name (6aR,9R)-4-(cyclopropanecarbonyl)-N,N-diethyl-7-methyl-6,6a,8,9-tetrahydroindolo[4,3-fg]quinoline-9-carboxamide
SMILES CCN(CC)C(=O)[C@H]1CN(C)[C@@H]2Cc3cn(C(=O)C4CC4)c5cccc(C2=C1)c35

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C24H29N3O2/c1-4-26(5-2)23(28)17-11-19-18-7-6-8-20-22(18)16(12-21(19)25(3)13-17)14-27(20)24(29)15-9-10-15/h6-8,11,14-15,17,21H,4-5,9-10,12-13H2,1-3H3/t17-,21-/m1/s1
Formula Weight 391.226
Chemical Name or Material 1CP-LSD (1-Cyclopropionyl lysergic acid diethylamide)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.