missing translation for 'onlineSavingsMsg'
Learn More

3,5,3',5'-Tetraiodo Thyromandelic Acid (>85%), TRC

Product Code. 30411212
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 30411212

Brand: Toronto Research Chemicals TRCT2961701MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3,5,3',5'-Tetraiodo Thyromandelic acid
CAS 93647-48-4
Molecular Formula C14 H8 I4 O5
Quantity 1 mg
Synonym Benzeneacetic acid','α-hydroxy-4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, acid','4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo-, 2-Hydroxy-2-[4-(4-hydroxy-3,5-diiodo-phenoxy)-3,5-diiodo-phenyl]acetic Acid, 3,5,3',5'-Tetraiodo Thyromandelic Acid, Mandelic acid','4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodo- (7CI), α-Hydroxy-4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodobenzeneacetic acid
InChI Formula InChI=1S/C14H8I4O5/c15-7-3-6(4-8(16)12(7)20)23-13-9(17)1-5(2-10(13)18)11(19)14(21)22/h1-4,11,19-20H,(H,21,22)
SMILES OC(C(=O)O)c1cc(I)c(Oc2cc(I)c(O)c(I)c2)c(I)c1
IUPAC Name 2-hydroxy-2-[4-(4-hydroxy-3,5-diiodophenoxy)-3,5-diiodophenyl]acetic acid
Molecular Weight (g/mol) 763.83
Formula Weight 763.655
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.